Merck
Reversal of RNA polymerase II ubiquitylation by the ubiquitin protease Ubp3.
Molecular cell (2008-05-24)
Kristian Kvint, Jay P Uhler, Michael J Taschner, Stefan Sigurdsson, Hediye Erdjument-Bromage, Paul Tempst, Jesper Q Svejstrup
RÉSUMÉ
The final outcome of protein polyubiquitylation is often proteasome-mediated proteolysis, meaning that "proofreading" of ubiquitylation by ubiquitin proteases (UBPs) is crucial. Transcriptional arrest can trigger ubiquitin-mediated proteolysis of RNA polymerase II (RNAPII) so a UBP reversing RNAPII ubiquitylation might be expected. Here, we show that Ubp3 deubiquitylates RNAPII in yeast. Genetic characterization of ubp3 cells is consistent with a role in elongation, and Ubp3 can be purified with RNAPII, Def1, and the elongation factors Spt5 and TFIIF. This Ubp3 complex deubiquitylates both mono- and polyubiquitylated RNAPII in vitro, and ubp3 cells have elevated levels of ubiquitylated RNAPII in vivo. Moreover, RNAPII is degraded faster in a ubp3 mutant after UV irradiation. Problems posed by damage-arrested RNAPII are thought to be resolved either by removing the damage or degrading the polymerase. In agreement with this, cells with compromised DNA repair are better equipped to survive UV damage when UPB3 is deleted.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
6-Azauracil, ≥98%