Merck
Oxidant-induced cell death in renal epithelial cells: differential effects of inorganic and organic hydroperoxides.
Pharmacology & toxicology (2003-04-25)
Sung Min Park, Hyun Chul Jung, Im Soo Koak, Ha Yun Na, Jae Suk Woo, Jin Sup Jung, Yong Keun Kim
RÉSUMÉ
This study was undertaken in order to examine the roles of lipid peroxidation and poly (ADP-ribose) polymerase (PARP) activation in oxidant-induced renal cell death. Opossum kidney cell cultures were used as the renal epithelial cell model, and an inorganic hydroperoxide H2O2 and an organic hydroperoxide t-butylhydroperoxide were employed as model oxidants. Cell death by both oxidants could be prevented by thiols (dithiothreitol and glutathione), iron chelators (deferoxamine and phenanthroline), and hydroxyl radical scavengers (dimethylthiourea and pyruvate). Phenolic antioxidants N,N'-diphenyl-p-phenylenediamine (DPPD) and butylated hydroxyanisole had no effect on the H2O2-induced cell death. However, the t-butylhydroperoxide-induced cell death was effectively prevented by these antioxidants. The PARP inhibitor 3-aminobenzamide prevented the cell death induced by H2O2, but not cell death by t-butylhydroperoxide. The PARP activity was increased in cells exposed to H2O2 but not t-butylhydroperoxide. Unlike in opossum kidney cells, in rabbit renal cortical slices both oxidants H2O2 and t-butylhydroperoxide induced cell death through a lipid peroxidation-dependent and PARP-independent mechanism. Effects of DPPD and 3-aminobenzamide on H2O2-induced cell death in primary cultured rabbit proximal tubular cells were similar to those in opossum kidney cells. These results indicate that 1) the H2O2-induced cell death in cultured renal epithelial cells is associated with PARP activation but not lipid peroxidation, whereas the t-butylhydroperoxide-induced cell death is mediated by lipid peroxidation, and 2) the role of lipid peroxidation in H2O2 cytotoxicity may be different between freshly isolated renal tubular cells and cultured renal epithelial cells.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
N,N′-Diphenyl-p-phenylenediamine, 98%