- Colorimetric method for the determination of lipoxygenase activity.
Colorimetric method for the determination of lipoxygenase activity.
Journal of agricultural and food chemistry (2001-02-15)
G E Anthon, D M Barrett
PMID11170556
RÉSUMÉ
A colorimetic assay for lipoxygenase activity has been developed. The assay is based on the detection of the lipoxygenase reaction product, linoleic acid hydroperoxide, by the oxidative coupling of 3-methyl-2-benzothiazolinone (MBTH) with 3-(dimethylamino)benzoic acid (DMAB) in a hemoglobin-catalyzed reaction. This test reaction is rapid and sensitive, and it offers advantages over other methods for detecting lipoxygenase activity. The assay is capable of detecting activity in a number of crude vegetable homogenates and should be particularly useful where a rapid visual determination of lipoxygenase activity is desired.