Merck
KLF8 promotes invasive outgrowth of breast cancer by inducing filopodium-like protrusions via CXCR4.
American journal of translational research (2022-03-12)
Debarati Mukherjee, Jie Hao, Heng Lu, Satadru K Lahiri, Lin Yu, Jihe Zhao
RÉSUMÉ
Post-therapeutic relapse remains the biggest challenge to breast cancer management. The re-initiation of proliferation of dormant tumor cells in either metastatic or primary tumor location marks the final rate-limiting step of malignancy and mortality. The underlying molecular mechanisms remain poorly understood. We have recently demonstrated that KLF8 promotes breast cancer metastasis via CXCR4 upregulation. Here we report a role and mechanisms for KLF8 in driving the recurrence-like tumor outgrowth in both secondary and primary sites in a CXCR4-dependent manner. Treatment of an MDA-MB-231 breast cancer cell variant with the CXCR4 ligand, CXCL12, induces formation of filopodia in monolayer culture and filopodium-like protrusions (FLPs) in 3D culture. The FLP+ cells proliferate significantly faster than FLP- cells in the 3D culture supplemented with CXCL12. Both the FLP formation and enhanced proliferation in the 3D culture can be prevented by silencing KLF8 expression in the cells. From this prevention, the cells can be rescued by overexpressing wild-type CXCR4 but not its inactive mutant form in the cells. Overexpression of KLF8 or CXCR4 in the cells dramatically enhances their invasive outgrowth and metastasis after being implanted into immunocompromised mice. Mechanistically, we found that the activated FAK was recruited to the nascent FLPs and that proliferation of the cells was completely prevented with a FAK-specific inhibitor. Taken together, these results shed new light on the role of KLF8 in promoting breast cancer recurrence, the fatal episode of the disease, by inducing CXCR4-dependent FLP formation.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
PF-573228, ≥95% (HPLC)