Merck
NF-kappaB is essential for epithelial-mesenchymal transition and metastasis in a model of breast cancer progression.
The Journal of clinical investigation (2004-08-18)
Margit A Huber, Ninel Azoitei, Bernd Baumann, Stefan Grünert, Andreas Sommer, Hubert Pehamberger, Norbert Kraut, Hartmut Beug, Thomas Wirth
RÉSUMÉ
The transcription factor NF-kappaB is activated in a range of human cancers and is thought to promote tumorigenesis, mainly due to its ability to protect transformed cells from apoptosis. To investigate the role of NF-kappaB in epithelial plasticity and metastasis, we utilized a well-characterized in vitro/in vivo model of mammary carcinogenesis that depends on the collaboration of the Ha-Ras oncoprotein and TGF-beta. We show here that the IKK-2/IkappaBalpha/NF-kappaB pathway is required for the induction and maintenance of epithelial-mesenchymal transition (EMT). Inhibition of NF-kappaB signaling prevented EMT in Ras-transformed epithelial cells, while activation of this pathway promoted the transition to a mesenchymal phenotype even in the absence of TGF-beta. Furthermore, inhibition of NF-kappaB activity in mesenchymal cells caused a reversal of EMT, suggesting that NF-kappaB is essential for both the induction and maintenance of EMT. In line with the importance of EMT for invasion, blocking of NF-kappaB activity abrogated the metastatic potential of mammary epithelial cells in a mouse model system. Collectively, these data provide evidence of an essential role for NF-kappaB during distinct steps of breast cancer progression and suggest that the cooperation of Ras- and TGF-beta-dependent signaling pathways in late-stage tumorigenesis depends critically on NF-kappaB activity.