Merck
Polysulfide Anions as Visible Light Photoredox Catalysts for Aryl Cross-Couplings.
Journal of the American Chemical Society (2020-12-29)
Haoyu Li, Xinxin Tang, Jia Hao Pang, Xiangyang Wu, Edwin K L Yeow, Jie Wu, Shunsuke Chiba
RÉSUMÉ
Polysulfide anions are endowed with unique redox properties, attracting considerable attentions for their applications in alkali metals-sulfur batteries. However, the employment of these anionic species in redox catalysis for small molecule synthesis remains underdeveloped due to their moderate-poor electrochemical potential in the ground state, whereas some of them are characterized by photoabsorptions in visible spectral regions. Herein, we disclose the use of polysulfide anions as visible light photoredox catalysts for aryl cross-coupling reactions. The reaction design enables single-electron reduction of aryl halides upon the photoexcitation of tetrasulfide dianions (S42-). The resulting aryl radicals are engaged in (hetero)biaryl cross-coupling, borylation, and hydrogenation in a redox catalytic regime involving S4• -/S42- and S3• -/S32- redox couples.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
1,1,2,2-Tetrachloroethane, reagent grade, ≥98%