Merck
  • Different effects of phospholipase Dζ2 and non-specific phospholipase C4 on lipid remodeling and root hair growth in Arabidopsis response to phosphate deficiency.
Different effects of phospholipase Dζ2 and non-specific phospholipase C4 on lipid remodeling and root hair growth in Arabidopsis response to phosphate deficiency.
The Plant journal : for cell and molecular biology (2018-02-14)
Yuan Su, Maoyin Li, Liang Guo, Xuemin Wang
RÉSUMÉ
Phosphate (Pi) deficiency in soils is a major limiting factor for plant growth. In response to Pi deprivation, one prominent metabolic adaptation in plants is the decrease in membrane phospholipids that consume approximately one-third cellular Pi. The level of two phospholipid-hydrolyzing enzymes, phospholipase Dζ2 (PLDζ2) and non-specific phospholipase C4 (NPC4), is highly induced in Pi-deprived Arabidopsis. To determine the role of PLDζ2 and NPC4 in plant growth under Pi limitation, Arabidopsis plants deficient in both PLDζ2 and NPC4 (npc4pldζ2) were generated and characterized. Lipid remodeling in leaves and roots was analyzed at three different durations of Pi deficiency. NPC4 affected lipid changes mainly in roots at an early stage of Pi deprivation, whereas PLDζ2 exhibited a more overt effect on lipid remodeling in leaves at a later stage of Pi deprivation. Pi deficiency-induced galactolipid increase and phospholipid decrease were impeded in pldζ2 and npc4pldζ2 plants. In addition, seedlings of npc4pldζ2 had the same root hair density as pldζ2 but shorter root hair length than pldζ2 in response to Pi deficiency. The loss of NPC4 decreased root hair length but had no effect on root hair density. These data suggest that PLDζ2 and NPC4 mediate the Pi deprivation-induced lipid remodeling in a tissue- and time-specific manner. PLDζ2 and NPC4 have distinctively different roles in root hair growth and development in response to Pi deprivation; PLDζ2 negatively modulates root hair density and length, whereas NPC4 promotes root hair elongation.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Murashige and Skoog Basal Salt Micronutrient Solution, 10 ×, liquid, suitable for plant cell culture