Merck
Mineralized Human Amniotic Membrane as a Biomimetic Scaffold for Hard Tissue Engineering Applications.
ACS biomaterials science & engineering (2021-01-16)
Leila Sabouri, Ali Farzin, Azadeh Kabiri, Peiman Brouki Milan, Mojtaba Farahbakhsh, Abolfazl Mehdizadehkashi, Abdolmohammad Kajbafzadeh, Ali Samadikuchaksaraei, Fatemeh Yousefbeyk, Mahmoud Azami, Mehdi Moghtadaei
RÉSUMÉ
The human amniotic membrane (HAM) has been viewed as a potential regenerative material for a wide variety of injured tissues because of its collagen-rich content. High degradability of HAM limits its wide practical application in bone tissue engineering. In this study, the natural matrix of the decellularized amniotic membrane was developed by the double diffusion method. The results confirmed a reduction of the amniotic membrane's degradability because of the deposition of calcium and phosphate ions during the double diffusion process. Real-time PCR results showed a high expression of osteogenesis-related genes from adipose-derived mesenchymal stem cells (ADMSCs) cultured on the surface of the developed mineralized amniotic membrane (MAM). Further in vivo experiments were conducted using an MAM preseeded with ADMSCs and a critical-size rat calvarial defect model. Histopathological results confirmed that the MAM + cell sample has excellent potential in bone regeneration.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Glutaraldéhyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative