Merck
Multiscale cardiac imaging spanning the whole heart and its internal cellular architecture in a small animal model.
eLife (2020-10-21)
Graham Rykiel, Claudia S López, Jessica L Riesterer, Ian Fries, Sanika Deosthali, Katherine Courchaine, Alina Maloyan, Kent Thornburg, Sandra Rugonyi
RÉSUMÉ
Cardiac pumping depends on the morphological structure of the heart, but also on its subcellular (ultrastructural) architecture, which enables cardiac contraction. In cases of congenital heart defects, localized ultrastructural disruptions that increase the risk of heart failure are only starting to be discovered. This is in part due to a lack of technologies that can image the three-dimensional (3D) heart structure, to assess malformations; and its ultrastructure, to assess organelle disruptions. We present here a multiscale, correlative imaging procedure that achieves high-resolution images of the whole heart, using 3D micro-computed tomography (micro-CT); and its ultrastructure, using 3D scanning electron microscopy (SEM). In a small animal model (chicken embryo), we achieved uniform fixation and staining of the whole heart, without losing ultrastructural preservation on the same sample, enabling correlative multiscale imaging. Our approach enables multiscale studies in models of congenital heart disease and beyond.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
L-Aspartic acid sodium salt monohydrate, ≥99.0% (NT)
Le tarif et la disponibilité ne sont pas disponibles actuellement.