Merck
  • Characterization and Cytotoxicity Analysis of a Ciprofloxacin Loaded Chitosan/Bioglass Scaffold on Cultured Human Periodontal Ligament Stem Cells: a Preliminary Report.
Characterization and Cytotoxicity Analysis of a Ciprofloxacin Loaded Chitosan/Bioglass Scaffold on Cultured Human Periodontal Ligament Stem Cells: a Preliminary Report.
Open access Macedonian journal of medical sciences (2016-10-06)
Maha I Abdelfattah, Sherine A Nasry, Amani A Mostafa
RÉSUMÉ
The aim of this study was to analyze the cytotoxicity of ciprofloxacin (CIP) loaded on chitosan bioactive glass scaffold on human periodontal ligament stem cells (PLSCs) in vitro. PLSCs obtained from human third molars, cultures treated with medium containing 15 x 15 mm chitosan/bioactive glass scaffolds without/with different concentration 0, 5, 10, and 20 % of CIP. A total of 15 x 10^3 cells were plated in 6 well plates. The attached cells of each group were harvested from the plates after 1, 4 and 8 days of culture to detect the viability of cells. The cell number was determined using a hemocytometer and the trypan blue dye-exclusion assay. Data was analyzed using normality using Shapiro-Wilk test. Comparisons between groups were made using One-way ANOVA complemented by Tukey's test. When comparing the proliferation rate of cells in the four groups, no statistically significant difference was found (P = 0.633). With regards to cell viability, no statistical difference was found between the 0, 5, and 10 % CIP concentrations, while the 20 % CIP concentration demonstrated the least viability with a high statistically significant difference (P = 0.003). Twenty percentages CIP demonstrated the least proliferation rate and viability.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-STRO-1 Antibody, clone STRO-1, ascites fluid, clone STRO-1, Chemicon®