Merck
Tropoelastin-Coated Tendon Biomimetic Scaffolds Promote Stem Cell Tenogenic Commitment and Deposition of Elastin-Rich Matrix.
ACS applied materials & interfaces (2019-05-16)
Helena Almeida, Rui M A Domingues, Suzanne M Mithieux, Ricardo A Pires, Ana I Gonçalves, Manuel Gómez-Florit, Rui L Reis, Anthony S Weiss, Manuela E Gomes
RÉSUMÉ
Tendon tissue engineering strategies that recreate the biophysical and biochemical native microenvironment have a greater potential to achieve regeneration. Here, we developed tendon biomimetic scaffolds using mechanically competent yarns of poly-ε-caprolactone, chitosan, and cellulose nanocrystals to recreate the inherent tendon hierarchy from a nano-to-macro scale. These were then coated with tropoelastin (TROPO) through polydopamine (PDA) linking, to mimic the native extracellular matrix (ECM) composition and elasticity. Both PDA and TROPO coatings decreased surface stiffness without masking the underlying substrate. We found that human adipose-derived stem cells (hASCs) seeded onto these TROPO biomimetic scaffolds more rapidly acquired their spindle-shape morphology and high aspect ratio characteristic of tenocytes. Immunocytochemistry shows that the PDA and TROPO-coated surfaces boosted differentiation of hASCs toward the tenogenic lineage, with sustained expression of the tendon-related markers scleraxis and tenomodulin up to 21 days of culture. Furthermore, these surfaces enabled the deposition of a tendon-like ECM, supported by the expression of collagens type I and III, tenascin, and decorin. Gene expression analysis revealed a downregulation of osteogenic and fibrosis markers in the presence of TROPO when compared with the control groups, suggesting proper ECM deposition. Remarkably, differentiated cells exposed to TROPO acquired an elastogenic profile due to the evident elastin synthesis and deposition, contributing to the formation of a more mimetic matrix in comparison with the PDA-coated and uncoated conditions. In summary, our biomimetic substrates combining biophysical and biological cues modulate stem cell behavior potentiating their long-term tenogenic commitment and the production of an elastin-rich ECM.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Monoclonal Anti-Elastin antibody produced in mouse, clone BA-4, ascites fluid