Merck
Postpartum supplementation with fermented ammoniated condensed whey altered nutrient partitioning to support hepatic metabolism.
Journal of dairy science (2020-06-15)
R Caputo Oliveira, S J Erb, R S Pralle, H T Holdorf, C R Seely, H M White
RÉSUMÉ
Our previously published paper demonstrated that fermented ammoniated condensed whey (FACW) supplementation improved feed efficiency and metabolic profile in postpartum dairy cows. The objective of this study was to further explore the effects of FACW supplementation on liver triglyceride content, hepatic gene expression and protein abundance, and plasma biomarkers related to liver function, inflammation, and damage. Individually fed multiparous Holstein cows were blocked by calving date and randomly assigned to postpartum (1 to 45 d in milk, DIM) isonitrogenous treatments: control diet (n = 20) or diet supplemented with FACW (2.9% dry matter of diet as GlucoBoost; Fermented Nutrition, Luxemburg, WI, replacing soybean meal; n = 19). Liver biopsies were performed at 14 and 28 DIM for analysis of mRNA expression, protein abundance, and liver triglyceride content. There was marginal evidence for a reduction in liver triglyceride content at 14 DIM in FACW-supplemented cows compared with the control group. Cows supplemented with FACW had greater mRNA expression of glucose-6-phosphatase at 14 DIM relative to control. Supplementation with FACW increased mRNA expression of pyruvate carboxylase (PC), but did not alter cytosolic phosphoenolpyruvate carboxykinase (PCK1), resulting in a 2.4-fold greater PC:PCK1 ratio for FACW-supplemented cows compared with control. There was no evidence for a FACW effect on mRNA expression of propionyl-CoA carboxylase nor on mRNA expression or protein abundance of lactate dehydrogenase A or B. Cows supplemented with FACW had lower plasma urea nitrogen compared with control. Plasma l-lactate was greater for FACW-supplemented cows compared with control at 2 h before feeding time at 21 DIM. There was no evidence for altered expression of IL1B or IL10, or blood biomarkers related to liver function and damage. Greater glucose-6-phosphatase and PC gene expression, together with greater blood glucose and similar milk lactose output, suggests that FACW increased the supply of glucose precursors, resulting in greater gluconeogenesis between 3 and 14 DIM. Greater hepatic PC:PCK1 ratio, together with previously reported decreased plasma β-hydroxybutyrate and the marginal evidence for lower liver triglyceride content at 14 DIM, suggests greater hepatic capacity for complete oxidation of fatty acids in FACW-supplemented cows compared with control. Overall, improvements in metabolite profile and feed efficiency observed with postpartum supplementation of FACW may be attributed to increased gluconeogenic and anaplerotic precursors, most likely propionate, due to modulated rumen fermentation.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
D-Lactate Colorimetric Assay, sufficient for 100 colorimetric tests