Merck
Evaluation of polyphenolic compounds in membrane concentrated pistachio hull extract.
Food chemistry (2018-12-07)
Negin Seifzadeh, Mohammad Ali Sahari, Mohsen Barzegar, Hassan Ahmadi Gavlighi, Luca Calani, Daniele Del Rio, Gianni Galaverna
RÉSUMÉ
Pistachio hull is a substantial source of natural polyphenols, but a substantial volume is being wasted annually. Aqueous pistachio green hull extract (PGHE) was subjected to two-stage membrane process in order to separate a polyphenol rich fraction. The membrane conditions of each stage were investigated. Total phenolic content and antioxidant activity were determined by Folin-Ciocalteu and DPPH· assay; also membrane fouling was monitored. The use of the 1 kDa cellulose membrane accompanied by 4 bar pressure and 250 rpm stirring speed was observed to be successful in the separation of a fraction with the highest amount of phenolic compound and antioxidant activity, in the retentate part. UHPLC/MSn characterization of PGHE polyphenols enabled us to identify 34 compounds, including the most abundant galloylshikimic acids, gallic acid, theogallin, galloyl-O-hexoside, quercetin-O-hexoside and pyrogallol.