Merck
  • Quantification of trans-resveratrol and its metabolites in human plasma using ultra-high performance liquid chromatography tandem quadrupole-orbitrap mass spectrometry.
Quantification of trans-resveratrol and its metabolites in human plasma using ultra-high performance liquid chromatography tandem quadrupole-orbitrap mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2018-11-20)
Ljubica Svilar, Jean-Charles Martin, Catherine Defoort, Charlotte Paut, Franck Tourniaire, Amandine Brochot
RÉSUMÉ
Trans-resveratrol is a stilbene polyphenol with a large spectrum of biological activities. This is why it is widely studied in terms of activities, bioavailability and quantitation in different foods, beverages and biological matrices. Different analytical methods are employed for its quantitation. In this study a quadrupole-orbitrap tandem mass spectrometer coupled to a reverse phase ultra-high performance liquid chromatography is applied to a quantitation of trans-resveratrol and its metabolites trans-resveratrol-3-O-β-d-glucuronide, trans-resveratrol-4'-O-β-d-glucuronide, trans-resveratrol-3-O-sulfate, a,b-dihydroresveratrol, a,b-dihydroresveratrol-glucuronide, a,b-dihydroresveratrol-glucuronide-sulfate, a,b-dihydroresveratrol-sulfate, trans-resveratrol-3,5-O-β-d-diglucuronide, trans-resveratrol-3,4'-O-d-β-diglucuronide, trans-resveratrol-3-O-β-d-glucuronide-sulfate and trans-resveratrol-4'-O-β-d-glucuronide-sulfate in human plasma. MS/MS experiments coupled to a high resolving power and accurate mass measurements as well as the use of labeled internal standards enabled the achievement of linear calibration curves across the four orders of magnitude concentration ranges. The method was validated in terms of specificity and selectivity, accuracy and precision, sensitivity and matrix effect and can be now applied to pharmacokinetic studies or routine analysis. In addition, the application of quadrupole-orbitrap mass spectrometer to the quantitation of trans-resveratrol and its metabolites provides acquisition of full collision induced dissociation spectra of analyzed compounds giving place to the structural characterization and sensitivity and linear concentration ranges respecting the accuracy and precision, specificity and selectivity requirements.