Merck
Efficient and Stable Inverted Planar Perovskite Solar Cells Using a Triphenylamine Hole-Transporting Material.
ChemSusChem (2018-04-08)
Rui Chen, Tongle Bu, Jing Li, Wei Li, Peng Zhou, Xueping Liu, Zhiliang Ku, Jie Zhong, Yong Peng, Fuzhi Huang, Yi-Bing Cheng, Zhengyi Fu
RÉSUMÉ
Inverted perovskite solar cells (PSCs) with a p-i-n structure have attracted great attention. Normally, inorganic p-type metal oxides or polymers are used as the hole-transport material (HTM), a vital component in the inverted PSCs. However, this type of HTM often requires high processing temperatures and/or high costs. On the other hand, a commonly used organic HTM, poly(3,4-ethylenedioxythiophene polystyrene sulfonate (PEDOT:PSS), is sensitive to humidity and thus affects the stability of the PSCs. Herein, we employ a small molecule, 4,4',4''-tris(N-3-methylphenyl-N-phenylamino) triphenylamine (m-MTDATA) to replace PEDOT:PSS as a new HTM for inverted PSCs. Compared to a PEDOT:PSS-based device, m-MTDATA-based PSCs exhibit enhanced performance. The highest power conversion efficiency (PCE) was notably improved from 13.44 % (PEDOT:PSS) to 18.12 % (m-MTDATA), suggesting that m-MTDATA could be an efficient HTM to achieve high performance inverted PSCs. Furthermore, the m-MTDATA-based device demonstrated improved stability (retaining 90 % PCE) under ambient conditions over 1000 h compared with the PEDOT:PSS-based devices (retaining 40 % PCE).