Merck
Purification of a 110-kDa phosphoinositide phospholipase C that is activated by G-protein beta gamma-subunits.
The Journal of biological chemistry (1993-11-25)
J L Blank, K Shaw, A H Ross, J H Exton
RÉSUMÉ
We report the purification from bovine brain cytosol of a 110-kDa phosphoinositide-specific phospholipase C (PLC-110) that was markedly stimulated by G-protein beta gamma-subunits. The enzyme was purified approximately 2000-fold with a yield of 4%. On the basis of size and immunological cross-reactivity, PLC-110 was distinct from 150-kDa PLC-beta 1, 145-kDa PLC-gamma 1, and 85-kDa PLC-delta 1. An antiserum to a peptide corresponding to a conserved PLC Y domain sequence cross-reacted with PLC-110. PLC-110 was also recognized by two antisera selective for NH2-terminal and internal sequences in PLC-beta 3, but not by a third peptide antiserum to the COOH terminus of this enzyme, suggesting that PLC-110 is related to PLC-beta 3. Reconstitution of purified PLC-110 with beta gamma-subunits produced greater than 100-fold activation, indicating activation was observed at approximately 60 nM beta gamma and full activation at approximately 500 nM beta gamma. PLC-110 maximally hydrolyzed phosphatidylinositol 4-phosphate and phosphatidylinositol 4,5-bisphosphate at 1 microM Ca2+, but showed no activity toward phosphatidylinositol at Ca2+ concentrations up to 1 mM. Concentrations of purified guanosine 5'-O-(3-thiotriphosphate)-liganded alpha q that fully activated PLC-beta 1 failed to stimulate PLC-110. This observation indicates that the site at which beta gamma interacts with PLC-110 is distinct from that at which alpha q regulates the activity of PLC-beta isozymes.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Phényl-Sepharose 6 Fast Flow (à débit élevé), low substitution, extent of labeling: ~20 μmol per mL