Merck
Neuroprotective effect of Ro5-4864 following brain injury.
Experimental neurology (2008-09-16)
Jean F Soustiel, Menashe Zaaroor, Eugene Vlodavsky, Leo Veenman, Abraham Weizman, Moshe Gavish
RÉSUMÉ
The 18 kDa translocator protein (TSPO) is a protein complex located at the outer mitochondrial membrane and interacting with the mitochondrial permeability transition pore (mPTP), indicating its involvement in the control of mPTP opening. We intended to explore the effect of TSPO ligands, PK 11195 and Ro5-4864 on apoptosis in a rat model of cortical injury. Sprague-Dawley rats received a daily intraperitoneal injection of dimethylsulfoxide (vehicle), PK 11195, or Ro5-4864, starting 2 days prior the injury and a third injection after the injury. At 6 weeks, immunohistochemistry analysis showed that Ro5-4864 resulted in a significant increase in the number of surviving neurons and in the density of the neurofilament network in the perilesional cortex in comparison with animals of the vehicle and PK 11195 groups. In tissue samples dissected from the injured area, Ro5-4864 caused a significant reduction in activation of caspases 3 and 9 but not of caspase 8 in comparison with the vehicle and PK 11195 groups. In addition, measurements of transmembrane mitochondrial potential of mitochondria (Deltapsi(M)) isolated from normal rat brain showed that loss of Deltapsi(M) induced by recombinant Bax could be significantly reduced by Ro5-4864 in a concentration-dependent manner. Our findings indicate that the neuroprotective effect shown by Ro5-4864 in the present model of brain injury involves the mitochondrial pathway of apoptosis modulation of mPTP.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Caspase 9 Colorimetric Activity Assay Kit, LEHD, Caspase-9 Colorimetric Activity Assay Kits provide a simple & convenient means for assaying the activity of caspases that recognize the LEHD.