Merck
Elongator mutation in mice induces neurodegeneration and ataxia-like behavior.
Nature communications (2018-08-12)
Marija Kojic, Monika Gaik, Bence Kiska, Anna Salerno-Kochan, Sarah Hunt, Angelo Tedoldi, Sergey Mureev, Alun Jones, Belinda Whittle, Laura A Genovesi, Christelle Adolphe, Darren L Brown, Jennifer L Stow, Kirill Alexandrov, Pankaj Sah, Sebastian Glatt, Brandon J Wainwright
RÉSUMÉ
Cerebellar ataxias are severe neurodegenerative disorders with an early onset and progressive and inexorable course of the disease. Here, we report a single point mutation in the gene encoding Elongator complex subunit 6 causing Purkinje neuron degeneration and an ataxia-like phenotype in the mutant wobbly mouse. This mutation destabilizes the complex and compromises its function in translation regulation, leading to protein misfolding, proteotoxic stress, and eventual neuronal death. In addition, we show that substantial microgliosis is triggered by the NLRP3 inflammasome pathway in the cerebellum and that blocking NLRP3 function in vivo significantly delays neuronal degeneration and the onset of ataxia in mutant animals. Our data provide a mechanistic insight into the pathophysiology of a cerebellar ataxia caused by an Elongator mutation, substantiating the increasing body of evidence that alterations of this complex are broadly implicated in the onset of a number of diverse neurological disorders.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
5-Methylcytidine, ≥99%