Merck
High resolution tissue mass spectrometry imaging reveals a refined functional anatomy of the human adult adrenal gland.
Endocrinology (2018-02-01)
Na Sun, Yin Wu, Kazutaka Nanba, Silviu Sbiera, Stefan Kircher, Thomas Kunzke, Michaela Aichler, Sabina Berezowska, Joachim Reibetanz, William E Rainey, Martin Fassnacht, Axel Walch, Matthias Kroiss
RÉSUMÉ
In the adrenal gland, neuroendocrine cells that synthesize catecholamines and epithelial cells that produce steroid hormones are united beneath a common organ capsule to function as a single stress-responsive organ. The functional anatomy of the steroid hormone producing adrenal cortex and the catecholamine producing medulla is ill defined at the level of small molecules. Here, we report the first comprehensive high-resolution mass spectrometry imaging (MSI) map of the normal human adrenal gland. A large variety of biomolecules was accessible by matrix-assisted laser desorption/ionization-Fourier transform-ion cyclotron resonance (MALDI-FT-ICR) MSI, including nucleoside phosphates indicative of oxidative phosphorylation, sterol and steroid metabolites, intermediates of glycolysis and the tricarboxylic acid (TCA) cycle, lipids and fatty acids. Statistical clustering analyses yielded a molecular defined adrenal anatomy of ten distinct molecular zones including a highly structured corticomedullary interface. By incorporating pathway information, activities of carbohydrate, amino acid and lipid metabolism as well as endocrine bioactivity were revealed to be highly spatially organized which could be visualized as different molecularly defined zones. Together, these findings provide a molecular definition of human adult adrenal gland structure beyond classical histological anatomy.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
9-Aminoacridine hydrochloride monohydrate, 98%