Merck
Increased extracellular clusterin in the prefrontal cortex in schizophrenia.
Schizophrenia research (2015-10-21)
Katina M Athanas, Sarah L Mauney, Tsung-Ung W Woo
RÉSUMÉ
The expression of the gene that encodes clusterin, a glycoprotein that has been implicated in the regulation of many cellular processes, has previously been found in gene expression profiling studies to be among the most significantly differentially expressed genes in pyramidal and parvalbumin-containing inhibitory neurons in the cerebral cortex in subjects with schizophrenia. In this study, we investigated whether clusterin may also be dysregulated at the protein level in schizophrenia subjects. We found that, although the intracellular amount of clusterin may be unchanged, the level of extracellular, secreted clusterin appears to be significantly increased in schizophrenia subjects. It is speculated that this finding may represent a neuroprotective response to pathophysiological events that underlie schizophrenia.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-Clusterin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution