Key Documents
Z381853
Ultramicro pipette tips
Microcapillary, flat tip, maximum volume 10 μL, non-sterile
About This Item
Code UNSPSC :
41121606
Nomenclature NACRES :
NB.01
Matériaux
colorless
polypropylene
polypropylene
Stérilité
non-sterile
Conditionnement
pack of 200 ea (Bulk packed)
Fabricant/nom de marque
Sorenson 1512
Volume maximum
10 μL
Description générale
Low capacity improves accuracy with small sample volumes by reducing dead space.
Application
For loading sample wells in 0.4 mm sequencing gels.
Autres remarques
Browse all pipette tips