Key Documents
Z177946
Scienceware® chemical spoons
L 21 cm
Synonyme(s) :
laboratory spatula, laboratory spoon
About This Item
Code UNSPSC :
41122403
Nomenclature NACRES :
NB.35
Matériaux
stainless steel
Fabricant/nom de marque
Bel-Art H367290021
Longueur
21 cm
Taille de l'extrémité en cuillère
1.6 mL , small
3.0 mL , large
Description générale
Double-ended with large-size spoon on one end and small spoon on the other. Solid, rounded connecting shaft.
Informations légales
Scienceware is a registered trademark of SP Scienceware, a division of SP