Key Documents
BR749311
BRAND® micro haematocrit capillary
sodium hep. w/o cal. mark I.D 1
Synonyme(s) :
Haematocrit capillaries
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.22
Matériaux
red
Caractéristiques
DIN ISO 12772 and BS 4316-68, To Contain
Conditionnement
pack of 1000 ea
Fabricant/nom de marque
BRAND 749311
Description générale
Compliant with DIN ISO 12772 and BS 4316-68 standards, these tubes feature a color code for easy identification and use in hematocrit centrifuges. The heparinized tubes are treated with sodium heparinate on the internal surface and are marked with a red color code, making them suitable for capillary blood samples. In contrast, the non-heparinized tubes are designated with a blue color code, specifically designed for use with heparinized venous blood.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
Certificats d'analyse (COA)
Lot/Batch Number
Désolés, nous n'avons pas de COA pour ce produit disponible en ligne pour le moment.
Les clients ont également consulté
Patrick Siyambulela Mafunda et al.
Animal reproduction science, 224, 106664-106664 (2020-12-05)
Several standard descriptions of the avian male and female reproductive tract have been reported, including effects of age, stage of reproductive maturity and gonadal hormone concentrations. Limited information on penguin reproductive biology and a lack of information on the African