Merck
  • Pulmonary injury associated with spray of a water-based nano-sized waterproofing product: a case study.
Pulmonary injury associated with spray of a water-based nano-sized waterproofing product: a case study.
Journal of occupational medicine and toxicology (London, England) (2017-12-14)
Paul T J Scheepers, Lucie Masen-Poos, Frits G B G J van Rooy, Arné Oerlemans, Eline van Daalen, Robbert Cremers, Hera Lichtenbeld, Bonne Biesma, Jorid B Sørli, Ismo K Koponen, Søren Thor Larsen, Peder Wolkoff, Asger W Nørgaard
ABSTRACT
In most reported cases of lung trauma with water proofing products, volatile organic compounds (VOC) have a prominent role. Here we report on a case involving ten workers exposed to a sprayed product containing nanoparticles in a water solution with only a few percent VOC. Ten workers suffered from respiratory symptoms following spray impregnation of hardwood furniture using a waterproofing product that contained positively charged fluorinated acrylate copolymer solid cores with a median diameter of 70 nm (1.3 w%) in aqueous suspension with 3.3 w% VOC and 0.3 w% quaternary ammonium. The worker who applied one liter of the product in a wood workshop, using an air mix spray gun, did not report any health complaints. Another worker, who entered the workshop 3 h later and had rolled and smoked two cigarettes, was hospitalized with severe chemical pneumonitis. A chest X-ray (CXR) showed bilateral infiltrative impairment in the lower lobe regions. On the next day a second CXR showed increased patchiness marking in all fields. A high-resolution Computer Tomography (CT)-scan demonstrated extensive bilateral areas of ground-glass opacities predominantly in the lower regions of the upper lobes, the right middle lobe and the apical regions of the lower lobes, compatible with severe chemical pneumonitis. On the following morning, nine workers in an adjacent workplace in the same building, experienced dry cough, chest tightness and substernal pain upon physical exercise. Reconstruction of the spray application in a climate chamber confirmed trimethyl silanol, glycol ethers and fluoroalkenes in the gas phase. Immediately after the spray application, aerosols were observed at a maximum concentration of 6.3 × 10 The hospitalized worker had smoked cigarettes contaminated with fluoropolymers which is a plausible explanation for the lung trauma. Respiratory symptoms in the nine workers may be caused by inhalation of particles that became airborne by resuspension from surfaces when workers entered the adjacent workplace the next day. A contribution from VOC appears less likely because measurements and modelling showed that concentrations in the mg/m
Sigma-Aldrich
1H,1H,2H,2H-Perfluoro-1-octanol, 97%
Sigma-Aldrich
Decamethyltetrasiloxane, 97%
Sigma-Aldrich
Trimethylsilanol, ≥97.5% (GC)
Sigma-Aldrich
1H,1H,2H,2H-Perfluorodecyl acrylate, contains 100 ppm tert-butylcatechol as inhibitor, 97%