Merck
  • Unveiling some FDA-approved drugs as inhibitors of the store-operated Ca
Unveiling some FDA-approved drugs as inhibitors of the store-operated Ca
Scientific reports (2017-10-19)
Saifur Rahman, Taufiq Rahman
ABSTRACT
The store-operated calcium entry (SOCE) pathway is an important route for generating cytosolic Ca
Sigma-Aldrich
1-Oleoyl-2-acetyl-sn-glycerol, ≥97% (TLC), oil