Merck
  • The pH-Triggered Triblock Nanocarrier Enabled Highly Efficient siRNA Delivery for Cancer Therapy.
The pH-Triggered Triblock Nanocarrier Enabled Highly Efficient siRNA Delivery for Cancer Therapy.
Theranostics (2017-09-16)
Lili Du, Junhui Zhou, Lingwei Meng, Xiaoxia Wang, Changrong Wang, Yuanyu Huang, Shuquan Zheng, Liandong Deng, Huiqing Cao, Zicai Liang, Anjie Dong, Qiang Cheng
ABSTRACT
Small interfering RNA (siRNA) therapies have been hampered by lack of delivery systems in the past decades. Nowadays, a few promising vehicles for siRNA delivery have been developed and it is gradually revealed that enhancing siRNA release from endosomes into cytosol is a very important factor for successful delivery. Here, we designed a novel pH-sensitive nanomicelle, PEG-PTTMA-P(GMA-S-DMA) (PTMS), for siRNA delivery. Owing to rapid hydrolysis in acidic environment, PTMS NPs underwent hydrophobic-to-hydrophilic transition in endosomes that enabled combination of proton sponge effect and raised osmotic pressure in endosomes, resulting in vigorous release of siRNAs from endosomes into cytosol.
Sigma-Aldrich
2,4,6-Trimethoxybenzaldehyde, 98%