Merck
  • Synthesis and characterization of 8-ethynyl-1,3-dihydro-benzo[b][1,4]diazepin-2-one derivatives: part 2. New potent non-competitive metabotropic glutamate receptor 2/3 antagonists.
Synthesis and characterization of 8-ethynyl-1,3-dihydro-benzo[b][1,4]diazepin-2-one derivatives: part 2. New potent non-competitive metabotropic glutamate receptor 2/3 antagonists.
Bioorganic & medicinal chemistry letters (2007-12-22)
Thomas J Woltering, Geo Adam, Jürgen Wichmann, Erwin Goetschi, James N C Kew, Frédéric Knoflach, Vincent Mutel, Silvia Gatti
ABSTRACT
A series of 1,3-dihydro-benzo[b][1,4]diazepin-2-one derivatives was evaluated as non-competitive mGluR2/3 antagonists. Replacement of a cyano group by a five-membered heterocycle produced compounds inhibiting the binding of [(3)H]-LY354740 to rat mGluR2 with low nanomolar affinity and consistent functional effect at both mGluR2 and mGluR3. Further modification to improve the physicochemical properties led eventually to compounds with the ability to reverse LY354740-mediated inhibition of field excitatory postsynaptic potentials in the rat dentate gyrus.
Product Description
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Discovery® Cyano Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4 mm