- Fate of the
Fate of the
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2016-11-08)
Karsten E Braun, Ann-Katrin Luks, Burkhard Schmidt
PMID27820683
ABSTRACT
The fate of
MATERIALS
The fate of