Merck
  • Fluconazole alters the polysaccharide capsule of Cryptococcus gattii and leads to distinct behaviors in murine Cryptococcosis.
Fluconazole alters the polysaccharide capsule of Cryptococcus gattii and leads to distinct behaviors in murine Cryptococcosis.
PloS one (2014-11-14)
Julliana Ribeiro Alves Santos, Rodrigo Assunção Holanda, Susana Frases, Mayara Bravim, Glauber de S Araujo, Patrícia Campi Santos, Marliete Carvalho Costa, Maira Juliana Andrade Ribeiro, Gabriella Freitas Ferreira, Ludmila Matos Baltazar, Aline Silva Miranda, Danilo Bretas Oliveira, Carolina Maria Araújo Santos, Alide Caroline Lima Fontes, Ludmila Ferreira Gouveia, Maria Aparecida Resende-Stoianoff, Jonatas Santos Abrahão, Antônio Lúcio Teixeira, Tatiane Alves Paixão, Danielle G Souza, Daniel Assis Santos
ABSTRACT
Cryptococcus gattii is an emergent human pathogen. Fluconazole is commonly used for treatment of cryptococcosis, but the emergence of less susceptible strains to this azole is a global problem and also the data regarding fluconazole-resistant cryptococcosis are scarce. We evaluate the influence of fluconazole on murine cryptococcosis and whether this azole alters the polysaccharide (PS) from cryptococcal cells. L27/01 strain of C. gattii was cultivated in high fluconazole concentrations and developed decreased drug susceptibility. This phenotype was named L27/01F, that was less virulent than L27/01 in mice. The physical, structural and electrophoretic properties of the PS capsule of L27/01F were altered by fluconazole. L27/01F presented lower antiphagocytic properties and reduced survival inside macrophages. The L27/01F did not affect the central nervous system, while the effect in brain caused by L27/01 strain began after only 12 hours. Mice infected with L27/01F presented lower production of the pro-inflammatory cytokines, with increased cellular recruitment in the lungs and severe pulmonary disease. The behavioral alterations were affected by L27/01, but no effects were detected after infection with L27/01F. Our results suggest that stress to fluconazole alters the capsule of C. gattii and influences the clinical manifestations of cryptococcosis.
Product Description
Amphotericin B, European Pharmacopoeia (EP) Reference Standard
Fluconazole for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)
Sigma-Aldrich
Fluconazole, ≥98% (HPLC), powder
Supelco
Fluconazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Heptane, biotech. grade, ≥99%
USP
Fluconazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Heptane, HPLC Plus, for HPLC, GC, and residue analysis, 99%
Sigma-Aldrich
Heptane, anhydrous, 99%
Sigma-Aldrich
Heptane, puriss. p.a., reag. Ph. Eur., ≥99% n-heptane basis (GC)
Supelco
Heptane, analytical standard
Levodopa, European Pharmacopoeia (EP) Reference Standard
USP
Levodopa, United States Pharmacopeia (USP) Reference Standard
Supelco
Levodopa, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Heptane, suitable for HPLC, ≥99%
Sigma-Aldrich
Heptane, suitable for HPLC, ≥96%
Sigma-Aldrich
Heptane, ReagentPlus®, 99%
Supelco
Heptane, Pharmaceutical Secondary Standard; Certified Reference Material
Fluconazole, European Pharmacopoeia (EP) Reference Standard