Merck
Xylitol as a prophylaxis for acute otitis media: systematic review.
International journal of audiology (2010-09-30)
Jeffrey L Danhauer, Carole E Johnson, Nicole E Corbin, Kaitlyn G Bruccheri
ABSTRACT
A systematic review was conducted to evaluate evidence regarding xylitol, a sugar alcohol, as a prophylaxis for acute otitis media (AOM) in children. The authors searched PubMed and other databases to identify evidence. Criteria for included studies were: appear in English-language, peer-reviewed journals; at least quasi-experimental designs; use xylitol; and present outcome data. The authors completed evaluation forms for the included studies at all phases of the review. The authors reviewed 1479 titles and excluded 1435. Abstracts and full texts were reviewed for the remaining 44; four randomized controlled trials met inclusion criteria. Xylitol was a generally well accepted prophylaxis for AOM with few side effects when administered via chewing gum or syrup at 10 g/day given five times daily. Meta-analysis revealed significant treatment effects (Risk ratio = 0.68; 95% confidence interval = 0.57 to 0.83). Xylitol can be a prophylaxis for AOM, but warrants further study, especially of vehicles other than chewing gum for young children, and information is needed regarding cost, duration of administration required, and expected long-term effects.
Product Description
Supelco
Xylitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Xylitol
Sigma-Aldrich
Xylitol, ≥99% (GC)
Xylitol, European Pharmacopoeia (EP) Reference Standard