Merck
  • [The use of sodium gamma-oxybutyrate and gutimine for decreasing metabolic disorders in the heart, caused by exotoxic shock in acetic acid poisoning].
[The use of sodium gamma-oxybutyrate and gutimine for decreasing metabolic disorders in the heart, caused by exotoxic shock in acetic acid poisoning].
Voprosy meditsinskoi khimii (1993-11-01)
I N Petrin, V T Dolgikh, I P Krolevets
ABSTRACT
Exotoxic shock was simulated in non-linear rat males anesthetized with barbital after intragastric administration of 70% acetic acid at a dose of 4 ml/kg using a gastric tube. Energy metabolism and the rate of lipid peroxidation were studied in heart muscle during the decompensated and terminal periods of exotoxic shock. Sodium hydroxybutyrate (200 mg/kg) and gutimine (75 mg/kg), administered alone within 30 min after the poisoning, prolonged the period before manifestation of the decompensated and terminal steps of the shock; these drugs decreased the rate of metabolic impairments in the myocardium by decreasing the rates of adenine nucleotide catabolism, of glycolysis, lipolysis and lipid peroxidation.
Sigma-Aldrich
2-Imino-4-thiobiuret, 99%