Merck
  • Calcium dobesilate reduces endothelin-1 and high-sensitivity C-reactive protein serum levels in patients with diabetic retinopathy.
Calcium dobesilate reduces endothelin-1 and high-sensitivity C-reactive protein serum levels in patients with diabetic retinopathy.
Molecular vision (2013-01-22)
Alireza Javadzadeh, Amir Ghorbanihaghjo, Farzad Hami Adl, Dima Andalib, Hassan Khojasteh-Jafari, Kamyar Ghabili
ABSTRACT
To determine the benefits of calcium dobesilate (CaD) administration on endothelial function and inflammatory status in patients with diabetic retinopathy through measurement of serum levels of endothelin-1 and high-sensitivity C-reactive protein (hsCRP). In a double-blind, randomized clinical trial, 90 patients with either severe nonproliferative or proliferative diabetic retinopathy and with blood glucose level of 120-200 mg/dl were randomly allocated to treatment with either CaD tablets (500 mg daily) or placebo for 3 months. Visual acuity, intraocular pressure, and macular status were performed before the study. The serum levels of endothelin-1 and hsCRP were evaluated in both groups before and at the third month of the trial. The median serum level of hsCRP significantly differed between the groups 3 months following the CaD or placebo administration (2.2 mg/l in the CaD group versus 3.7 mg/l in the placebo group, p=0.01). The mean endothelin-1 serum level was 0.69±0.32 pg/ml in the CaD group and 0.86±0.30 pg/ml in the placebo group (p=0.01). Furthermore, in the CaD group, the serum levels of both endothelin-1 and hsCRP were significantly decreased 3 months after administration of CaD (p<0.001). Administration of the CaD in the patients with diabetic retinopathy may reduce the serum levels of endothelin-1 and hsCRP. This might imply amelioration of the endothelial function and inflammatory status following CaD therapy in these patients.
Sigma-Aldrich
Dobesilate Calcium, ≥98% (HPLC)
Sigma-Aldrich
Hydroquinonesulfonic acid potassium salt, technical grade