Merck
  • Enhanced oral bioavailability and anti-tumour effect of paclitaxel by 20(s)-ginsenoside Rg3 in vivo.
Enhanced oral bioavailability and anti-tumour effect of paclitaxel by 20(s)-ginsenoside Rg3 in vivo.
Biopharmaceutics & drug disposition (2012-08-18)
Lei-Qiong Yang, Bin Wang, Hui Gan, Shou-Ting Fu, Xiao-Xia Zhu, Zhuo-Na Wu, Da-Wei Zhan, Ruo-Lan Gu, Gui-Fang Dou, Zhi-Yun Meng
ABSTRACT
The purpose of this study was to investigate the effect of paclitaxel in combination with 20(s)-ginsenoside Rg3 on its anti-tumour effect in nude mice. In the Caco-2 transport assay, the apparent permeability from the apical side to the basal side (P(app)) (A-B) and P(app) (B-A) of paclitaxel were measured when co-incubated with different concentrations of 20(s)-ginsenoside Rg3. The results indicated that the penetration of paclitaxel through the Caco-2 monolayer from the apical side to the basal side was facilitated by 20(s)-ginsenoside Rg3 in a concentration-dependent manner. Meanwhile, 20(s)-ginsenoside Rg3 inhibited P-glycoprotein (P-gp), and the maximum inhibition was achieved at 80 µM (p < 0.05). The pharmacokinetic parameters of paclitaxel after oral co-administration of paclitaxel (40 mg/kg) with various doses of 20(s)-ginsenoside Rg3 in rats were investigated by an in vivo pharmacokinetic experiment. The results showed that the AUC of paclitaxel co-administered with 20(s)-ginsenoside Rg3 was significantly higher (p < 0.001 at 10 mg/kg) compared with the control. The relative bioavailability (RB) % of paclitaxel with 20(s)-ginsenoside Rg3 was 3.4-fold (10 mg/kg) higher than that of the control. The effect of paclitaxel orally co-administered with 20(s)-ginsenoside Rg3 against human tumour MCF-7 xenografts in nude mice was also evaluated. Paclitaxel (20 mg/kg) co-administered with 20(s)-ginsenoside Rg3 (10 mg/kg) exhibited an effective anti-tumour activity with the relative tumor growth rate (T/C) values of 39.36% (p <0.05). The results showed that 20(s)-ginsenoside Rg3 enhanced the oral bioavailability of paclitaxel in rats and improved the anti-tumour activity in nude mice, indicating that oral co-administration of paclitaxel with 20(s)-ginsenoside Rg3 could provide an effective strategy in addition to the established i.v. route.
Sigma-Aldrich
Ginsenoside Rg3, ≥98% (HPLC)
Supelco
Ginsenoside Rg3, analytical standard