- KCa channels and epoxyeicosatrienoic acids: major contributors to thermal hyperaemia in human skin.
KCa channels and epoxyeicosatrienoic acids: major contributors to thermal hyperaemia in human skin.
While it is accepted that NO is responsible for ā¼60% of the plateau in cutaneous thermal hyperaemia, a large portion of the response remains unknown. We sought to determine whether the remaining ā¼40% could be attributed to EDHF-mediated activation of KCa channels, and whether the epoxyeicosatrienoic acids (EETs), derived via cytochrome P450, were the predominant EDHF active in the response. Four microdialysis fibres were placed in the forearm skin of 20 subjects. In Protocol 1 (n = 10): (1) Control, (2) N(G)-nitro-l-arginine methyl ester (l-NAME), (3) a KCa channel inhibitor, tetraethylammonium (TEA), and (4) TEA + l-NAME. In Protocol 2 (n = 10): (1) Control, (2) l-NAME, (3) a cytochrome P450 inhibitor, sulfaphenazole, and (4) sulfaphenazole + l-NAME. Local heating to 42Ā°C was performed and skin blood flow was measured with laser Doppler flowmetry. Data are presented as the percentage of maximal cutaneous vascular conductance (CVC). All drug sites attenuated plateau CVC from the control site (86 Ā± 1%) to 79 Ā± 3% with sulfaphenazole (P = 0.02 from control), 71 Ā± 3% with TEA (P = 0.01 from control), and further to 38 Ā± 2% with l-NAME (P < 0.001 from control, P < 0.001 from TEA). Plateau was largely attenuated with sulfaphenazole + l-NAME (24 Ā± 2%; P = 0.002 from l-NAME), and nearly abolished with l-NAME + TEA (13 Ā± 2%; P = 0.001 from sulfaphenazole + l-NAME), which was not different from baseline (P = 0.14). Furthermore, the initial peak was just 17 Ā± 2% with TEA + l-NAME (P < 0.001 from l-NAME). These data suggest EDHFs are responsible for a large portion of initial peak and the remaining 40% of the plateau phase, as administration of TEA in combination with l-NAME abolished the majority of hyperaemia. These data also suggest EETs contribute to about half of the EDHF response.