Merck
  • Fast preparation of photopolymerized poly(benzyl methacrylate-co-bisphenol A dimethacrylate) monoliths for capillary electrochromatography.
Fast preparation of photopolymerized poly(benzyl methacrylate-co-bisphenol A dimethacrylate) monoliths for capillary electrochromatography.
Journal of chromatography. A (2010-04-17)
Junjie Ou, Graham T T Gibson, Richard D Oleschuk
ABSTRACT
A novel porous polymer monolith was prepared in situ in a fused-silica capillary using photoinitiated polymerization. Bisphenol A dimethacrylate (BPADMA) was selected as a crosslinker, copolymerized with benzyl methacrylate (BMA) in the presence of a binary porogenic solvent consisting of cyclohexanol and 1-decanol in < or =10 min. The resulting poly(BMA-co-BPADMA) monoliths exhibited good permeability and mechanical stability. Mixtures of alkylbenzenes, polycyclic aromatic hydrocarbons (PAHs) or phenolic compounds were successfully separated by CEC. A similar monolith was also prepared with ethylene dimethacrylate (EDMA) as the crosslinker instead of BPADMA to compare the separation ability of the resulting monoliths. The results indicated that poly(BMA-co-BPADMA) monoliths have better selectivity for aromatic analytes and greater chromatographic stability in higher aqueous mobile phase.
Sigma-Aldrich
Bisphenol A dimethacrylate, >98%