Merck
  • Capillary electrophoresis of chitooligosaccharides in acidic solution: simple determination using a quaternary-ammonium-modified column and indirect photometric detection with crystal violet.
Capillary electrophoresis of chitooligosaccharides in acidic solution: simple determination using a quaternary-ammonium-modified column and indirect photometric detection with crystal violet.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2009-12-17)
Toshiaki Hattori, Nobuhiro Anraku, Ryo Kato
ABSTRACT
Five chitosan oligosaccharides were separated in acidic aqueous solution by capillary electrophoresis (CE) with indirect photometric detection using a positively coated capillary. Electrophoretic mobility of the chitooligosaccharides (COSs) depended on the number of monomer units in acidic aqueous solution, similar to other polyelectrolyte oligomers. The separation was developed in nitric acid aqueous solution at pH 3.0 with 1 mM Crystal Violet, using a capillary positively coated with N-trimethoxypropyl-N,N,N-trimethylammonium chloride. The limit of the detection for chitooligosaccharides with two to six saccharide chains was less than 5 microM. CE determination of an enzymatically hydrolyzed COS agreed with results from HPLC.
Sigma-Aldrich
Tetraphenylphosphonium bromide, 97%
Sigma-Aldrich
Tetraphenylphosphonium chloride, 98%