Merck
  • Design and synthesis of hydrophobic and chiral anions from amino acids as precursor for functional ionic liquids.
Design and synthesis of hydrophobic and chiral anions from amino acids as precursor for functional ionic liquids.
Chemical communications (Cambridge, England) (2006-07-21)
Kenta Fukumoto, Hiroyuki Ohno
ABSTRACT
Hydrophobic ionic liquids composed of tetrabutylphosphonium cation and chiral anions derived from amino acids modified with trifluoromethane sulfonyl groups have been synthesized using a simple method.
Sigma-Aldrich
Tetrabutylphosphonium chloride, 96%
Sigma-Aldrich
Tetrabutylphosphonium bromide, 98%