Merck
  • N-t-Butyl hydroxylamine regulates heat shock-induced apoptosis in U937 cells.
N-t-Butyl hydroxylamine regulates heat shock-induced apoptosis in U937 cells.
Redox report : communications in free radical research (2006-01-28)
Hyun Jeong Kim, Seoung Woo Shin, Chang Joo Oh, Mann Hyung Lee, Chae Ha Yang, Jeen-Woo Park
ABSTRACT
Heat shock may increase oxidative stress due to increased production of reactive oxygen species and/or the promotion of cellular oxidation events. Therefore, compounds that scavenge reactive oxygen species may regulate heat shock-induced cell death. Recently, it has been shown that the decomposition product of the spin-trapping agent alpha-phenyl-N-t-butylnitrone, N-t-butyl hydroxylamine (NtBHA), mimics alpha-phenyl-N-t-butylnitrone and is much more potent in delaying reactive oxygen species-associated senescence. We investigated the protective role of NtBHA against heat shock-induced apoptosis in U937 cells. Upon exposure to heat shock, there was a distinct difference between the untreated cells and the cells pre-treated with 0.1 mM NtBHA for 2 h in regard to apoptotic parameters, cellular redox status, and mitochondrial function. Upon exposure to heat shock, NtBHA pre-treated cells showed significant inhibition of apoptotic features such as activation of caspase-3, up-regulation of Bax, and down-regulation of Bcl-2 compared to untreated cells. This study indicates that NtBHA may play an important role in regulating the apoptosis induced by heat shock, presumably through scavenging of reactive oxygen species.
Sigma-Aldrich
N-tert-Butylhydroxylamine hydrochloride, ≥98%