Merck
  • Potassium nitrite reaction with 11-nor-delta 9-tetrahydrocannabinol-9-carboxylic acid in urine in relation to the drug screening analysis.
Potassium nitrite reaction with 11-nor-delta 9-tetrahydrocannabinol-9-carboxylic acid in urine in relation to the drug screening analysis.
Journal of forensic sciences (1999-09-16)
S A Lewis, L A Lewis, A Tuinman
ABSTRACT
Recently potassium nitrite has been used as an adulterant to interfere with the analysis of 11-nor-delta 9-tetrahydro-cannabinol-9-carboxylic acid (THC-COOH) in urine. A comprehensive study of the THC-COOH and nitrite reaction chemistry and stability under various conditions is presented. Reverse phase high performance liquid chromatography (HPLC) and negative electrospray mass spectrometry (ESMS) results are given to substantiate the derived reaction mechanism and properties leading to reaction termination. The addition of potassium carbonate as a buffering agent prior to or following sample void as a means of preventing the formation of a nitroso-complexed form of the 11-nor-delta 9+-tetrahydrocannabinol-9-carboxylic acid is evaluated.
Sigma-Aldrich
Potassium carbonate, meets USP testing specifications
Sigma-Aldrich
Potassium nitrite, meets analytical specification of FCC, 97-100.5%
Sigma-Aldrich
Potassium nitrite, ACS reagent, ≥96.0%