Merck
  • Pseudouridine synthases modify human pre-mRNA co-transcriptionally and affect pre-mRNA processing.
Pseudouridine synthases modify human pre-mRNA co-transcriptionally and affect pre-mRNA processing.
Molecular cell (2022-01-21)
Nicole M Martinez, Amanda Su, Margaret C Burns, Julia K Nussbacher, Cassandra Schaening, Shashank Sathe, Gene W Yeo, Wendy V Gilbert
ABSTRACT
Pseudouridine is a modified nucleotide that is prevalent in human mRNAs and is dynamically regulated. Here, we investigate when in their life cycle mRNAs become pseudouridylated to illuminate the potential regulatory functions of endogenous mRNA pseudouridylation. Using single-nucleotide resolution pseudouridine profiling on chromatin-associated RNA from human cells, we identified pseudouridines in nascent pre-mRNA at locations associated with alternatively spliced regions, enriched near splice sites, and overlapping hundreds of binding sites for RNA-binding proteins. In vitro splicing assays establish a direct effect of individual endogenous pre-mRNA pseudouridines on splicing efficiency. We validate hundreds of pre-mRNA sites as direct targets of distinct pseudouridine synthases and show that PUS1, PUS7, and RPUSD4-three pre-mRNA-modifying pseudouridine synthases with tissue-specific expression-control widespread changes in alternative pre-mRNA splicing and 3' end processing. Our results establish a vast potential for cotranscriptional pre-mRNA pseudouridylation to regulate human gene expression via alternative pre-mRNA processing.
Sigma-Aldrich
Anti-U1-70K Antibody, clone 9C4.1, clone 9C4.1, from mouse
Sigma-Aldrich
Rosetta 2(DE3)pLysS Competent Cells - Novagen, Novagen′s Rosetta 2 (pLysS) host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli along with T7 lysozyme activity.
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution