Merck
Progerin modulates the IGF-1R/Akt signaling involved in aging.
Science advances (2022-07-21)
Bo Jiang, Xuan Wu, Fang Meng, Limiao Si, Sunrun Cao, Yuqing Dong, Huayi Sun, Mengzhu Lv, Hongde Xu, Ning Bai, Qiqiang Guo, Xiaoyu Song, Yang Yu, Wendong Guo, Fei Yi, Tingting Zhou, Xiaoman Li, Yanling Feng, Zhuo Wang, Dan Zhang, Yi Guan, Mengtao Ma, Jingwei Liu, Xining Li, Weidong Zhao, Baohua Liu, Toren Finkel, Liu Cao
ABSTRACT
Progerin, a product of LMNA mutation, leads to multiple nuclear abnormalities in patients with Hutchinson-Gilford progeria syndrome (HGPS), a devastating premature aging disorder. Progerin also accumulates during physiological aging. Here, we demonstrate that impaired insulin-like growth factor 1 receptor (IGF-1R)/Akt signaling pathway results in severe growth retardation and premature aging in Zmpste24-/- mice, a mouse model of progeria. Mechanistically, progerin mislocalizes outside of the nucleus, interacts with the IGF-1R, and down-regulates its expression, leading to inhibited mitochondrial respiration, retarded cell growth, and accelerated cellular senescence. Pharmacological treatment with the PTEN (phosphatase and tensin homolog deleted on chromosome 10) inhibitor bpV (HOpic) increases Akt activity and improves multiple abnormalities in Zmpste24-deficient mice. These findings provide previously unidentified insights into the role of progerin in regulating the IGF-1R/Akt signaling in HGPS and might be useful for treating LMNA-associated progeroid disorders.
Sigma-Aldrich
Anti-prelamin A Antibody, clone PL-1C7, clone PL-1C7, from mouse