Merck
  • Glioblastoma stem cell-specific histamine secretion drives pro-angiogenic tumor microenvironment remodeling.
Glioblastoma stem cell-specific histamine secretion drives pro-angiogenic tumor microenvironment remodeling.
Cell stem cell (2022-10-21)
Jiayi Chen, Guangqin Liu, Xinzheng Wang, Hao Hong, Tingting Li, Lin Li, Hongxiang Wang, Jiong Xie, Bohan Li, Ting Li, Dingyi Lu, Yakun Zhang, Haixin Zhao, Chengcheng Yao, Kaiqing Wen, Teng Li, Jing Chen, Shengming Wu, Kun He, Wei-Na Zhang, Jie Zhao, Na Wang, Qiuying Han, Qing Xia, Ji Qi, Juxiang Chen, Tao Zhou, Jianghong Man, Xue-Min Zhang, Ai-Ling Li, Xin Pan
ABSTRACT
The communication between glioblastoma stem cells (GSCs) and the surrounding microenvironment is a prominent feature accounting for the aggressive biology of glioblastoma multiforme (GBM). However, the mechanisms by which GSCs proactively drive interactions with microenvironment is not well understood. In this study, we interrogated metabolites that are preferentially secreted from GSCs and found that GSCs produce and secrete histamine to shape a pro-angiogenic tumor microenvironment. This histamine-producing ability is attributed to H3K4me3 modification-activated histidine decarboxylase (HDC) transcription via MYC. Notably, HDC is highly expressed in GBM, which is associated with poor survival of these patients. GSC-secreted histamine activates endothelial cells by triggering a histamine H1 receptor (H1R)-Ca2+-NF-κB axis, thereby promoting angiogenesis and GBM progression. Importantly, pharmacological blockage of H1R using antihistamines impedes the growth of GBM xenografts in mice. Our findings establish that GSC-specific metabolite secretion remodels the tumor microenvironment and highlight histamine targeting as a potential strategy for GBM therapy.
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit
Sigma-Aldrich
ChIPAb+ Acetyl-Histone H3 (Lys27) - ChIP Validated Antibody and Primer Set, clone CMA309, from mouse
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Diphenhydramine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Anti-monomethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit