Merck
  • Augmented lipopolysaccharide-induced TNF-alpha production by peritoneal macrophages in type 2 diabetic mice is dependent on elevated glucose and requires p38 MAPK.
Augmented lipopolysaccharide-induced TNF-alpha production by peritoneal macrophages in type 2 diabetic mice is dependent on elevated glucose and requires p38 MAPK.
Journal of immunology (Baltimore, Md. : 1950) (2007-01-05)
Christina L Sherry, Jason C O'Connor, Jason M Kramer, Gregory G Freund
ABSTRACT
Dysregulated inflammation is a complication of type 2 diabetes (T2D). In this study, we show that augmented LPS-induced TNF-alpha production by resident peritoneal macrophages (PerMphi) in type 2 diabetic (db/db) mice is dependent on elevated glucose and requires p38 MAPK. Intraperitoneal LPS administered to db/db and nondiabetic (db/+) mice induced 3- and 4-fold more TNF-alpha in the peritoneum and serum, respectively, of db/db mice as compared with db/+ mice. Examination of the TLR-4/MD2 complex and CD14 expression showed no difference between db/db and db/+ PerMphi. Ex vivo stimulation of PerMphi with LPS produced a similar 3-fold increase in TNF-alpha production in db/db PerMphi when compared with db/+ PerMphi. PerMphi isolated from db/+ mice incubated in high glucose (4 g/L) medium for 12 h produced nearly 2-fold more TNF-alpha in response to LPS than PerMphi incubated in normal glucose medium (1 g/L). LPS-dependent stimulation of PI3K activity, ERK1/2 activation, and p38 kinase activity was greater in PerMphi from db/db mice as compared with db/+ mice. Only inhibition of p38 kinase blocked LPS-induced TNF-alpha production in PerMphi from db/db mice. Taken together, these data indicate that augmented TNF-alpha production induced by LPS in macrophages during diabetes is due to hyperglycemia and increased LPS-dependent activation of p38 kinase.
Sigma-Aldrich
PD 98059, PD 98059, CAS 167869-21-8, is a cell-permeable, selective & reversible inhibitor of MAP Kinase Kinase (MEK). Inhibits MAP Kinase activation and subsequent phosphorylation of MAP Kinase substrates.
Millipore
SB 202190, SB 202190, CAS 152121-30-7, is a potent, reversible, competitive inhibitor of p38. Inhibits p38 phosphorylation of myelin basic protein. Blocks the activity of p38β (Ki = 16 nM; IC₅₀ = 350 nM).
Sigma-Aldrich
LY 294002, InSolution, ≥98%, 10 mM, reversible and specific inhibitor of PI 3-kinase