Merck
  • Adult bone marrow is a rich source of human mesenchymal 'stem' cells but umbilical cord and mobilized adult blood are not.
Adult bone marrow is a rich source of human mesenchymal 'stem' cells but umbilical cord and mobilized adult blood are not.
British journal of haematology (2003-04-16)
Sarah A Wexler, Craig Donaldson, Patricia Denning-Kendall, Claire Rice, Ben Bradley, Jill M Hows
ABSTRACT
In postnatal life, mesenchymal stem cells (MSC) self-replicate, proliferate and differentiate into mesenchymal tissues, including bone, fat, tendon, muscle and bone marrow (BM) stroma. Possible clinical applications for MSC in stem cell transplantation have been proposed. We have evaluated the frequency, phenotype and differentiation potential of MSC in adult BM, cord blood (CB) and peripheral blood stem cell collections (PBSC). During culture, BM MSC proliferated to confluence in 10-14 d, maintaining a stable non-haemopoietic phenotype, HLA class-1+, CD29+, CD44+, CD90+, CD45-, CD34- and CD14 through subsequent passages. Using the colony forming unit fibroblasts assay, the estimated frequency of MSC in the BM nucleated cell population was 1 in 3.4 x 10(4) cells. Both adipogenic and osteogenic differentiation of BM MSC was demonstrated. In contrast, CB and PBSC mononuclear cells cultured in MSC conditions for two passages produced a population of adherent, non-confluent fibroblast-like cells with a haemopoietic phenotype, CD45+, CD14+, CD34-, CD44-, CD90- and CD29-. In paired experiments, cultured BM MSC and mature BM stroma were seeded with CB cells enriched for CD34+. Similar numbers of colony-forming units of granulocytes-macrophages were produced by MSC-based and standard stroma cultures over 10 weeks. We conclude that adult BM is a reliable source of functional cultured MSC, but CB and PBSC are not.
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Millipore
MultiScreen-PCRµ96 Filter Plate, Purification of PCR products
Sigma-Aldrich
Hanks′ Balanced Salt solution, With sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - low glucose, With 1000 mg/L glucose and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture