Merck
Serotonin Expression in the Song Circuitry of Adult Male Zebra Finches.
Neuroscience (2020-06-27)
Ping K Yip, Magdalena Schmitzberger, Mohammed Al-Hasan, Julia George, Elina Tripoliti, Adina T Michael-Titus, David Clayton, John V Priestley
ABSTRACT
Serotonin is an important neurotransmitter of the brain, but its role in song control remains to be fully demonstrated. Using male zebra finches (Taeniopygia guttata) that have song learning and production capabilities, we analysed the serotonin expression levels in the song nuclei and adjacent areas (peri-song nuclei) using immunohistochemistry. Key song nuclei were identified using combinations of Hoechst, choline acetyltransferase, and a neurofilament (NN18) marker in reference to the ZEBrA atlas. Mean serotonin expression was highest in interfacial nucleus (Nif) and lower in the other song nuclei in the following order (in order of highest first): interfacial nucleus (Nif) > Area X > dorsomedial part of the intercollicular nucelus (DM) > robust nucleus of the archistriatum (RA) > lateral magnocellular nucleus of the anterior neostriatum (LMAN) > ventral respiratory group (VRG) > dorsolateral nucleus of the medial thalamus (DLM) > the nucleus HVC (proper name) > tracheosyringeal motor nucleus (nXIIts). However, the mean serotonin expression (in order of highest first) in the peri-song nuclei regions was: peri-DM > peri-nXIIts > supra-peri-HVC > peri-RA > peri-DLM > peri-Area X > infra-peri-HVC > peri-VRG > peri-LMAN > peri-Nif. Interestingly, serotoninergic fibers immunostained for serotonin or the serotonin transporter can be found as a basket-like peri-neuronal structure surrounding cholinergic cell bodies, and appear to form contacts onto dopaminergic neurones. In summary, serotonin fibers are present at discrete song nuclei, and peri-song nuclei regions, which suggest serotonin may have a direct and/or modulatory role in song control.
Sigma-Aldrich
Anti-Tyrosine Hydroxylase antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution, sufficient for 10 blots
Sigma-Aldrich
Anti-Neurofilament 160 kDa Antibody, clone NN18, clone NN18, Chemicon®, from mouse
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat