Merck
  • N-benzoylated phenoxazines and phenothiazines: synthesis, antiproliferative activity, and inhibition of tubulin polymerization.
N-benzoylated phenoxazines and phenothiazines: synthesis, antiproliferative activity, and inhibition of tubulin polymerization.
Journal of medicinal chemistry (2011-05-14)
Helge Prinz, Behfar Chamasmani, Kirsten Vogel, Konrad J Böhm, Babette Aicher, Matthias Gerlach, Eckhard G Günther, Peter Amon, Igor Ivanov, Klaus Müller
ABSTRACT
A total of 53 N-benzoylated phenoxazines and phenothiazines, including their S-oxidized analogues, were synthesized and evaluated for antiproliferative activity, interaction with tubulin, and cell cycle effects. Potent inhibitors of multiple cancer cell lines emerged with the 10-(4-methoxybenzoyl)-10H-phenoxazine-3-carbonitrile (33b, IC(50) values in the range of 2-15 nM) and the isovanillic analogue 33c. Seventeen compounds strongly inhibited tubulin polymerization with activities higher than or comparable to those of the reference compounds such as colchicine. Concentration-dependent flow cytometric studies revealed that inhibition of K562 cell growth was associated with an arrest in the G2/M phases of the cell cycle, indicative of mitotic blockade. Structure-activity relationship studies showed that best potencies were obtained with agents bearing a methoxy group placed para at the terminal phenyl ring and a 3-cyano group in the phenoxazine. A series of analogues highlight not only the phenoxazine but also the phenothiazine structural scaffold as valuable pharmacophores for potent tubulin polymerization inhibitors, worthy of further investigation.
Sigma-Aldrich
Vinblastine sulfate salt, ≥97% (HPLC)
Sigma-Aldrich
Phenoxazine, 97%
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
2-Chlorophenothiazine, 97%
Sigma-Aldrich
Phenothiazine, purum, ≥98.0% (GC)
Sigma-Aldrich
Phenothiazine, ≥98%
Sigma-Aldrich
Podophyllotoxin
Sigma-Aldrich
Colchicine, BioReagent, suitable for plant cell culture, ≥95% (HPLC)
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder