Merck
  • A fluorescence-microscopic and cytofluorometric system for monitoring the turnover of the autophagic substrate p62/SQSTM1.
A fluorescence-microscopic and cytofluorometric system for monitoring the turnover of the autophagic substrate p62/SQSTM1.
Autophagy (2011-04-05)
Amena BenYounès, Nicolas Tajeddine, Maximilien Tailler, Shoaib Ahmad Malik, Shensi Shen, Didier Métivier, Oliver Kepp, Ilio Vitale, Maria Chiara Maiuri, Guido Kroemer
ABSTRACT
Autophagic flux can be measured by determining the declining abundance of autophagic substrates such as sequestosome 1 (SQSTM1, better known as p62), which is sequestered in autophagosomes upon its direct interaction with LC3. However, the total amount of p62 results from two opposed processes, namely its synthesis (which can be modulated by some cellular stressors including autophagy inducers) and its degradation. To avoid this problem, we generated a stable cell line expressing a chimeric protein composed by p62 and the HaloTag (®) protein, which serves as a receptor for fluorescent HaloTag (®) ligands. Upon labeling with HaloTag (®) ligands (which form covalent, near-to-undissociable bonds with the Halotag (®) receptor) and washing, the resulting fluorescent labeling is not influenced by de novo protein synthesis, therefore allowing for the specific monitoring of the fusion protein decline without any interference by protein synthesis. We demonstrate that a HaloTag (®) -p62 fusion protein stably expressed in suitable cell lines can be used to monitor autophagy by flow cytometry and automated fluorescence microscopy. We surmise that this system could be adapted to high-throughput applications.
Sigma-Aldrich
Pifithrin-α, A cell-permeable chemical inhibitor of p53.
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®