Merck
Microscaled proteogenomic methods for precision oncology.
Nature communications (2020-01-29)
Shankha Satpathy, Eric J Jaehnig, Karsten Krug, Beom-Jun Kim, Alexander B Saltzman, Doug W Chan, Kimberly R Holloway, Meenakshi Anurag, Chen Huang, Purba Singh, Ari Gao, Noel Namai, Yongchao Dou, Bo Wen, Suhas V Vasaikar, David Mutch, Mark A Watson, Cynthia Ma, Foluso O Ademuyiwa, Mothaffar F Rimawi, Rachel Schiff, Jeremy Hoog, Samuel Jacobs, Anna Malovannaya, Terry Hyslop, Karl R Clauser, D R Mani, Charles M Perou, George Miles, Bing Zhang, Michael A Gillette, Steven A Carr, Matthew J Ellis
ABSTRACT
Cancer proteogenomics promises new insights into cancer biology and treatment efficacy by integrating genomics, transcriptomics and protein profiling including modifications by mass spectrometry (MS). A critical limitation is sample input requirements that exceed many sources of clinically important material. Here we report a proteogenomics approach for core biopsies using tissue-sparing specimen processing and microscaled proteomics. As a demonstration, we analyze core needle biopsies from ERBB2 positive breast cancers before and 48-72 h after initiating neoadjuvant trastuzumab-based chemotherapy. We show greater suppression of ERBB2 protein and both ERBB2 and mTOR target phosphosite levels in cases associated with pathological complete response, and identify potential causes of treatment resistance including the absence of ERBB2 amplification, insufficient ERBB2 activity for therapeutic sensitivity despite ERBB2 amplification, and candidate resistance mechanisms including androgen receptor signaling, mucin overexpression and an inactive immune microenvironment. The clinical utility and discovery potential of proteogenomics at biopsy-scale warrants further investigation.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Aprotinin Nicotiana tobacco, >= 5TIU/mg protein, >= 98 % SDS-PAGE | 9087-70-1, recombinant, expressed in Nicotiana (tobacco), ≥5 TIU/mg protein, ≥98% (SDS-PAGE)