Merck
Transthyretin expression in the postischemic brain.
PloS one (2019-09-04)
Daniela Talhada, Isabel Gonçalves, João Carlos Gomes, Maria João Saraiva, Cecília Reis Santos, Karsten Ruscher
ABSTRACT
The unknown role of the carrier protein transthyretin (TTR) in mechanisms of functional recovery in the postischemic brain prompted us to study its expression following experimental stroke. Male C57/B6 mice (age 9 to 10 weeks) were subjected to permanent focal ischemia induced by photothrombosis (PT) and brain tissues were analyzed for ttr expression and TTR levels at 24 hours, 48 hours, 7 days and 14 days following the insult by RT-PCR, Western blot and immunohistochemistry. Fourteen days after PT, non-specific TTR-like immunoreactive globules were found in the ischemic core and surrounding peri-infarct region by immunohistochemistry that could not be allocated to DAPI positive cells. No TTR immunoreactivity was found when stainings were performed with markers for neurons (Neuronal Nuclei, NeuN), reactive astrocytes (glial fibrillary acidic protein, GFAP) or microglia (cluster of differentiation 68, CD68). In addition, we could not find TTR by immunoblotting in protein extracts obtained from the ischemic territory nor ttr expression by RT-PCR at all time points following PT. In all experiments, ttr expression in the choroid plexus and TTR in the mouse serum served as positive controls and recombinant legumain peptide as negative control. Together, our results indicate that TTR is not synthesized in brain resident cells in the ischemic infarct core and adjacent peri-infarct area. Thus, it seems unlikely that in situ synthesized TTR is involved in mechanisms of tissue reorganization during the first 14 days following PT.
Sigma-Aldrich
Transthyretin Blocking Peptide for SAB3500378
Sigma-Aldrich
Anti-Transthyretin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution