Merck
  • Identification of serotonin 2A receptor as a novel HCV entry factor by a chemical biology strategy.
Identification of serotonin 2A receptor as a novel HCV entry factor by a chemical biology strategy.
Protein & cell (2018-03-16)
Lin Cao, Jizheng Chen, Yaxin Wang, Yuting Yang, Jie Qing, Zihe Rao, Xinwen Chen, Zhiyong Lou
ABSTRACT
Hepatitis C virus (HCV) is a leading cause of liver disease worldwide. Although several HCV protease/polymerase inhibitors were recently approved by U.S. FDA, the combination of antivirals targeting multiple processes of HCV lifecycle would optimize anti-HCV therapy and against potential drug-resistance. Viral entry is an essential target step for antiviral development, but FDA-approved HCV entry inhibitor remains exclusive. Here we identify serotonin 2A receptor (5-HT2AR) is a HCV entry factor amendable to therapeutic intervention by a chemical biology strategy. The silencing of 5-HT2AR and clinically available 5-HT2AR antagonist suppress cell culture-derived HCV (HCVcc) in different liver cells and primary human hepatocytes at late endocytosis process. The mechanism is related to regulate the correct plasma membrane localization of claudin 1 (CLDN1). Moreover, phenoxybenzamine (PBZ), an FDA-approved 5-HT2AR antagonist, inhibits all major HCV genotypes in vitro and displays synergy in combination with clinical used anti-HCV drugs. The impact of PBZ on HCV genotype 2a is documented in immune-competent humanized transgenic mice. Our results not only expand the understanding of HCV entry, but also present a promising target for the invention of HCV entry inhibitor.
Sigma-Aldrich
SB 699551 dihydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Anti-Claudin 1 antibody produced in rabbit, affinity isolated antibody